accessibility menu, dialog, popup

UserName

ClNCS

CAS RN 2740-81-0

IUPAC Name: 1-chloro-2-isothiocyanatobenzene
Synonyms: 1-chloro-2-isothiocyanatobenzene
Molecular Weight (g/mol): 169.63
Molecular Formula: C7H4ClNS
InChi Key: DASSPOJBUMBXLU-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1N=C=S
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results