missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenyl isothiocyanate, 97%

Product Code. 11493767
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493767 25 g 25g
11483767 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493767 Supplier Thermo Scientific Alfa Aesar Supplier No. A18238.14
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2740-81-0 | C7H4ClNS | 169.626 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2740-81-0
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.626
MDL Number MFCD00004801
InChI Key DASSPOJBUMBXLU-UHFFFAOYSA-N
Synonym 2-chlorophenyl isothiocyanate, 2-chlorophenylisothiocyanate, benzene, 1-chloro-2-isothiocyanato, benzene, chloroisothiocyanato, 1-chloro-2-isothiocyanato-benzene, isothiocyanic acid 2-chlorophenyl ester, 2-chlorobenzenisothiocyanate, acmc-1ccw5, o-chlorophenylisothiocyanate, o-chlorophenyl isothiocyanate
PubChem CID 123171
IUPAC Name 1-chloro-2-isothiocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=S)Cl

Specifications

Density 1.287
Quantity 25 g
Odor Pungent
Refractive Index 1.6608
UN Number UN2922
Beilstein 2081473
Boiling Point 260°C to 262°C
Formula Weight 169.64
Percent Purity 97%
Chemical Name or Material 2-Chlorophenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.