accessibility menu, dialog, popup

UserName

CH3SOOHO

CAS RN 27489-33-4

IUPAC Name: 2-methanesulfonylphenol
Synonyms: 2-methanesulfonylphenol
Molecular Weight (g/mol): 172.2
Molecular Formula: C7H8O3S
InChi Key: ZWTZHHHSRDUVRT-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results