accessibility menu, dialog, popup

UserName

HOONHHN

CAS RN 2762-32-5

IUPAC Name: piperazine-2-carboxylic acid
Synonyms: piperazine-2-carboxylic acid
Molecular Weight (g/mol): 130.15
Molecular Formula: C5H10N2O2
InChi Key: JSSXHAMIXJGYCS-UHFFFAOYNA-N
SMILES: OC(=O)C1CNCCN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results