accessibility menu, dialog, popup

UserName

OHPHOHO

CAS RN 2767-80-8

IUPAC Name: [bis(hydroxymethyl)phosphanyl]methanol
Synonyms: methanol, phosphinidynetris-
Molecular Weight (g/mol): 124.08
Molecular Formula: C3H9O3P
InChi Key: JMXMXKRNIYCNRV-UHFFFAOYSA-N
SMILES: OCP(CO)CO
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)