missing translation for 'onlineSavingsMsg'
Learn More

Tris(hydroxymethyl)phosphine, 95%

Product Code. 10262952 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10262952 1 g Glass bottle 1g
10465375 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10262952 Supplier Thermo Scientific Acros Supplier No. 363840010
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2767-80-8 | C3H9O3P | 124.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2767-80-8
Molecular Formula C3H9O3P
Molecular Weight (g/mol) 124.08
MDL Number MFCD00055382
InChI Key JMXMXKRNIYCNRV-UHFFFAOYSA-N
Synonym tris hydroxymethyl phosphine, methanol, phosphinidynetris, trimethylolphosphine, phosphinidynetrimethanol, tris methanol phosphine, phosphinidynetrismethanol, unii-y6tg7wf7oq, y6tg7wf7oq, methanol, 1,1',1-phosphinidynetris, methanol, phosphinidynetri-7ci,8ci
PubChem CID 76001
IUPAC Name bis(hydroxymethyl)phosphanylmethanol
SMILES C(O)P(CO)CO

Specifications

Melting Point 48°C to 56°C
Color White to Yellow
Boiling Point 111.0°C to 113.0°C (2.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 124.08
Percent Purity 95%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Tris(hydroxymethyl)phosphine
Show More Show Less
compliance-icons
Product Identifier
  • Tris(hydroxymethyl)phosphine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.