accessibility menu, dialog, popup

UserName

CAS RN 27935-87-1

I

CAS RN 27935-87-1

IUPAC Name: (iodomethyl)cyclopentane
Synonyms: (iodomethyl)cyclopentane
Molecular Weight (g/mol): 210.06
Molecular Formula: C6H11I
InChi Key: DUMSKQUKLVSSII-UHFFFAOYSA-N
SMILES: ICC1CCCC1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)