missing translation for 'onlineSavingsMsg'
Learn More

(Iodomethyl)cyclopentane, 98+%, stab. with copper

Product Code. 11491010
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491010 5 g 5g
11401020 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491010 Supplier Thermo Scientific Alfa Aesar Supplier No. B25391.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 27935-87-1 | C6H11I | 210.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 27935-87-1
Molecular Formula C6H11I
Molecular Weight (g/mol) 210.06
MDL Number MFCD03453190
InChI Key DUMSKQUKLVSSII-UHFFFAOYSA-N
Synonym iodomethyl cyclopentane, cyclopentylmethyl iodide, cyclopentane, iodomethyl, cyclopentyliodomethane, acmc-20akpz, cyclopentylmethyliodide, iodomethyl-cyclopentane, cyclopentanemethyl iodide
PubChem CID 2762531
IUPAC Name iodomethylcyclopentane
SMILES ICC1CCCC1

Spezifikation

Density 1.614
Boiling Point 78°C (17 mmHg)
Flash Point 92°C (198°F)
Refractive Index 1.542
Quantity 5 g
Sensitivity Light Sensitive
Formula Weight 210.06
Percent Purity ≥98%
Chemical Name or Material (Iodomethyl)cyclopentane, stabilized with copper

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.