accessibility menu, dialog, popup

UserName

H3COBrOH

CAS RN 28165-49-3

IUPAC Name: 2-bromo-6-methoxyphenol
Synonyms: 2-bromo-6-methoxyphenol
Molecular Weight (g/mol): 203.04
Molecular Formula: C7H7BrO2
InChi Key: WEUFQISIJPSTBM-UHFFFAOYSA-N
SMILES: COC1=C(O)C(Br)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results