accessibility menu, dialog, popup

UserName

CAS RN 28178-42-9

IUPAC Name: 2-isocyanato-1,3-bis(propan-2-yl)benzene
Synonyms: 2-isocyanato-1,3-diisopropylbenzene
Molecular Weight (g/mol): 203.29
Molecular Formula: C13H17NO
InChi Key: FEUFNKALUGDEMQ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C(C)C)=C1N=C=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results