accessibility menu, dialog, popup

UserName

CAS RN 2829-58-5

NH2NSNHOO

CAS RN 2829-58-5

IUPAC Name: 4-amino-1,2,5-thiadiazole-3-carboxylic acid
Synonyms: 4-amino-1,2,5-thiadiazole-3-carboxylic acid
Molecular Weight (g/mol): 145.14
Molecular Formula: C3H3N3O2S
InChi Key: OJMKAQZLZNNVTG-UHFFFAOYSA-N
SMILES: NC1=NSN=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results