missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-[1,2,5]thiadiazole-3-carboxylic acid, Thermo Scientific™

Product Code. 10259424
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10259424 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10259424 Supplier Thermo Scientific Maybridge Supplier No. MAY00178DA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 2829-58-5
Packaging Amber Glass Bottle
Molecular Formula C3H3N3O2S
MDL Number MFCD00718830
Quantity 1 g
Synonym 4-amino-1,2,5 thiadiazole-3-carboxylic acid, 4-amino-1,2,5-thiadiazole-3-carboxylicacid, 1,2,5-thiadiazole-3-carboxylic acid, 4-amino, 1,2,5-thiadiazole, tda2, 1,2,5-thiadiazole-3-carboxylicacid, 4-amino, 4-amino-1,2,5-thiadiazol-3-carboxylic acid
InChI Key OJMKAQZLZNNVTG-UHFFFAOYSA-N
SMILES NC1=NSN=C1C(O)=O
IUPAC Name 4-amino-1,2,5-thiadiazole-3-carboxylic acid
Molecular Weight (g/mol) 145.14
PubChem CID 251763
Formula Weight 145.14
Percent Purity 97%
Chemical Name or Material 4-Amino-[1, 2, 5]thiadiazole-3-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.