accessibility menu, dialog, popup

UserName

CH3HOH2N

CAS RN 2835-97-4

IUPAC Name: 2-amino-3-methylphenol
Synonyms: 2-amino-3-methylphenol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: FEDLEBCVFZMHBP-UHFFFAOYSA-N
SMILES: CC1=CC=CC(O)=C1N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)