accessibility menu, dialog, popup

UserName

CH3NH2OOH

CAS RN 2840-04-2

IUPAC Name: 5-amino-2-methylbenzoic acid
Synonyms: 5-amino-2-methylbenzoic acid
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: FSXVZWAWYKMFMX-UHFFFAOYSA-N
SMILES: CC1=C(C=C(N)C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)