missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-methylbenzoic Acid, TRC
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
missing translation for 'unitSize'
25g
Specifications
Specifications
| Chemical Name or Material | 5-Amino-2-methylbenzoic Acid |
| CAS | 2840-04-2 |
| Molecular Formula | C8H9NO2 |
| Quantity | 25 g |
| Synonym | 3-Amino-6-methylbenzoic Acid, 5-Amino-o-toluic Acid |
| InChI Formula | InChI=1S/C8H9NO2/c1-5-2-3-6(9)4-7(5)8(10)11/h2-4H,9H2,1H3,(H,10,11) |
| SMILES | Cc1ccc(N)cc1C(=O)O |
| IUPAC Name | 5-amino-2-methylbenzoic acid |
| Molecular Weight (g/mol) | 151.16 |
| Formula Weight | 151.0633 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction