accessibility menu, dialog, popup

UserName

CAS RN 28556-81-2

IUPAC Name: 2-isocyanato-1,3-dimethylbenzene
Synonyms: 2-isocyanato-1,3-dimethylbenzene
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: YQLRKXVEALTVCZ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1N=C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results