missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenyl isocyanate, 98%

Product Code. 11353376
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353376 25 g 25g
11333376 1 g 1g
11343376 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11353376 Supplier Thermo Scientific Alfa Aesar Supplier No. L12805.14
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 28556-81-2 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28556-81-2
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00002012
InChI Key YQLRKXVEALTVCZ-UHFFFAOYSA-N
Synonym 2,6-dimethylphenyl isocyanate, benzene, 2-isocyanato-1,3-dimethyl, 2,6-dimethylphenylisocyanate, 2,6-xylyl isocyanate, unii-2ql5mu0e0c, 2ql5mu0e0c, isocyanic acid 2,6-dimethylphenyl ester, 2,6-dimethylbenzenisocyanate, 2,6-dimethyl phenyl isocyanate, 2-isocyanato-1,3-dimethyl-benzene
PubChem CID 98787
IUPAC Name 2-isocyanato-1,3-dimethylbenzene
SMILES CC1=CC=CC(C)=C1N=C=O

Specifications

Density 1.057
Quantity 25 g
Refractive Index 1.5356
UN Number UN2206
Beilstein 742593
Boiling Point 71°C to 72°C (3 mmHg)
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material 2,6-Dimethylphenyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.