accessibility menu, dialog, popup

UserName

S

CAS RN 287-27-4

IUPAC Name: thietane
Synonyms: thietane
Molecular Weight (g/mol): 74.14
Molecular Formula: C3H6S
InChi Key: XSROQCDVUIHRSI-UHFFFAOYSA-N
SMILES: C1CSC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results