missing translation for 'onlineSavingsMsg'
Learn More

Trimethylene sulfide

Product Code. 16594194 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16594194 25 g 25g
16584194 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16594194 Supplier Thermo Scientific Acros Supplier No. 182910250
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 287-27-4 | C3H6S | 74.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 287-27-4
Molecular Formula C3H6S
Molecular Weight (g/mol) 74.14
MDL Number MFCD00042751
InChI Key XSROQCDVUIHRSI-UHFFFAOYSA-N
IUPAC Name thietane
SMILES C1CSC1

Specifications

Density 1.0280 g/mL
Boiling Point 94.0°C to 94.5°C
Flash Point -2°C
Refractive Index 1.5040 to 1.5140
Quantity 25 g
Beilstein 17, I, 3
Specific Gravity 1.028
Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 74.14
Chemical Name or Material Trimethylene sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.