accessibility menu, dialog, popup

UserName

CAS RN 2873-97-4

CH3OCH3H3CNHOH2C

CAS RN 2873-97-4

IUPAC Name: N-(2-methyl-4-oxopentan-2-yl)prop-2-enamide
Synonyms: diacetone acrylamide
Molecular Weight (g/mol): 169.22
Molecular Formula: C9H15NO2
InChi Key: OMNKZBIFPJNNIO-UHFFFAOYSA-N
SMILES: CC(=O)CC(C)(C)NC(=O)C=C
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)