missing translation for 'onlineSavingsMsg'
Learn More

Diacetone acrylamide, 99%, ACROS Organics™

Product Code. 10623162 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10623162

Brand: Thermo Scientific Acros 352320050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2873-97-4
Melting Point 53°C to 57°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H15NO2
Linear Formula CH2CHC(O)NHC(CH3)2CH2C(O)CH3
MDL Number MFCD00008788
Quantity 5 g
Synonym diacetone acrylamide, diacetoneacrylamide, n-1,1-dimethyl-3-oxobutyl acrylamide, 2-propenamide, n-1,1-dimethyl-3-oxobutyl, n-2-methyl-4-oxopentan-2-yl acrylamide, n-2-2-methyl-4-oxopentyl acrylamide, acrylamide, n-1,1-dimethyl-3-oxobutyl, n-1,1-dimethyl-3-oxobutyl-2-propenamide, acrylamide, n,n-diacetonyl, ccris 5898
InChI Key OMNKZBIFPJNNIO-UHFFFAOYSA-N
SMILES CC(=O)CC(C)(C)NC(=O)C=C
IUPAC Name N-(2-methyl-4-oxopentan-2-yl)prop-2-enamide
Molecular Weight (g/mol) 169.22
PubChem CID 17888
Formula Weight 169.23
Percent Purity ≥98.5%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Diacetone acrylamide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Diacetone acrylamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.