accessibility menu, dialog, popup

UserName

H3COHNSOOHNH2HCl

CAS RN 28798-28-9

IUPAC Name: 2-amino-3-[(acetamidomethyl)sulfanyl]propanoic acid hydrochloride
Synonyms: 2-amino-3-[(acetamidomethyl)sulfanyl]propanoic acid hydrochloride
Molecular Weight (g/mol): 228.69
Molecular Formula: C6H13ClN2O3S
InChi Key: SZWPOAKLKGUXDD-UHFFFAOYNA-N
SMILES: Cl.CC(=O)NCSCC(N)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results