missing translation for 'onlineSavingsMsg'
Learn More

S-Acetamidomethyl-L-cysteine hydrochloride, 98%, Thermo Scientific™

Product Code. 15449718
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449718 5 g 5g
15438166 250 mg 250mg
15439718 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15449718 Supplier Thermo Scientific Alfa Aesar Supplier No. H63171.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28798-28-9
Molecular Formula C6H13ClN2O3S
Molecular Weight (g/mol) 228.691
MDL Number MFCD00077080
InChI Key SZWPOAKLKGUXDD-JEDNCBNOSA-N
Synonym h-cys acm-oh hcl, s-acetamidomethyl-l-cysteine hydrochloride, h-cys acm-oh.hcl, 2r-2-amino-3-acetamidomethyl sulfanyl propanoic acid hydrochloride, l-cysteine, s-acetylamino methyl-, monohydrochloride, h-cys acm .hcl, h-cys acm-oh hydrochloride, s-acetamidomethylcysteine hydrochloride, s-acetyl-aminomethyl-l-cysteine hydrochloride, h-cys acm-oh inverted exclamation mark currencyhcl
PubChem CID 16211002
IUPAC Name (2R)-3-(acetamidomethylsulfanyl)-2-aminopropanoic acid;hydrochloride
SMILES CC(=O)NCSCC(C(=O)O)N.Cl

Specifications

Melting Point 165°C (decomposition)
Beilstein 4015821
Quantity 5 g
Formula Weight 228.7
Percent Purity 98%
Physical Form Solid
Chemical Name or Material S-Acetamidomethyl-L-cysteine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.