accessibility menu, dialog, popup

UserName

CAS RN 288-36-8

HNNN

CAS RN 288-36-8

IUPAC Name: 2H-1,2,3-triazole
Synonyms: 2H-1,2,3-triazole osotriazole
Molecular Weight (g/mol): 69.07
Molecular Formula: C2H3N3
InChi Key: QWENRTYMTSOGBR-UHFFFAOYSA-N
SMILES: N1N=CC=N1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)