missing translation for 'onlineSavingsMsg'
Learn More

1H-1,2,3-Triazole, TRC

Product Code. 30454235
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30454235 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454235 Supplier Toronto Research Chemicals Supplier No. TRCT7673901G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 288-36-8
Molecular Formula C2H3N3
Quantity 1 g
Synonym v-Triazole (7CI,8CI), 1,2,3-Triazole, 2,3-Diazapyrrole, 2H-1,2,3-Triazole, Osotriazole, Pyrrodiazole, Triazacyclopentadiene
InChI Formula InChI=1S/C2H3N3/c1-2-4-5-3-1/h1-2H,(H,3,4,5)
SMILES c1c[nH]nn1
IUPAC Name 1H-triazole
Molecular Weight (g/mol) 69.07
Formula Weight 69.0327
Chemical Name or Material 1H-1,2,3-Triazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.