accessibility menu, dialog, popup

UserName

CAS RN 2888-06-4

IUPAC Name: 3-chlorobenzene-1-sulfonyl chloride
Synonyms: 3-chlorobenzenesulfonyl chloride
Molecular Weight (g/mol): 211.06
Molecular Formula: C6H4Cl2O2S
InChi Key: OINWZUJVEXUHCC-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)S(Cl)(=O)=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)