missing translation for 'onlineSavingsMsg'
Learn More

3-chlorobenzenesulfonyl chloride, 98%

Product Code. 10219593 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10219593 25 g Glass bottle 25g
10539882 1 g Glass bottle 1g
10056093 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10219593 Supplier Thermo Scientific Acros Supplier No. 356480250
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2888-06-4 | C6H4Cl2O2S | 211.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2888-06-4
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.07
MDL Number MFCD00051697
InChI Key OINWZUJVEXUHCC-UHFFFAOYSA-N
Synonym 3-chlorobenzene-1-sulfonyl chloride, 3-chlorobenzenesulphonyl chloride, 3-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro, m-chlorobenzenesulfonyl chloride, 3-chloro-benzenesulfonyl chloride, 3-chlorophenylsulfonyl chloride, pubchem5076, m-chlorbenzolsulfochlorid, acmc-1cj4b
PubChem CID 17909
IUPAC Name 3-chlorobenzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)Cl)S(=O)(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.4990g/mL
Boiling Point 102°C (0.9 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClC6H4SO2Cl
Refractive Index 1.5670 to 1.569
Quantity 25 g
Specific Gravity 1.499
Formula Weight 211.07
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-chlorobenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chlorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.