CAS RN 289472-78-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-Hydroxy Des(boric Acid) Bortezomib, TRC
CAS: 289472-78-2 Molecular Formula: C19 H24 N4 O3 Molecular Weight (g/mol): 356.42 Synonym: 2-Pyrazinecarboxamide, N-[(1S)-2-[[(1R)-1-hydroxy-3-methylbutyl]amino]-2-oxo-1-(phenylmethyl)ethyl]-,Pyrazinecarboxamide, N-[(1S)-2-[[(1R)-1-hydroxy-3-methylbutyl]amino]-2-oxo-1-(phenylmethyl)ethyl]- (9CI),N-[(1S)-2-[[(1R)-1-Hydroxy-3-methylbutyl]amino]-2-oxo-1-(phenylmethyl)ethyl]-2-pyrazinecarboxamide,(S,R)-Bortezomib hydroxyisopentyl amide analog (USP) IUPAC Name: N-[(2S)-1-[[(1R)-1-hydroxy-3-methylbutyl]amino]-1-oxo-3-phenylpropan-2-yl]pyrazine-2-carboxamide SMILES: CC(C)C[C@@H](O)NC(=O)[C@H](Cc1ccccc1)NC(=O)c2cnccn2