CAS RN 290821-39-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cimiracemoside D, MedChemExpress
MedChemExpress Cimiracemoside D is a natural product found in Actaea racemosa with unknown details.