accessibility menu, dialog, popup

UserName

CAS RN 2909-77-5

IUPAC Name: N,N-dimethyl-2,6-bis(propan-2-yl)aniline
Synonyms: 2,6-diisopropyl-N,N-dimethylaniline
Molecular Weight (g/mol): 205.35
Molecular Formula: C14H23N
InChi Key: ALXIOUGHHXXLKX-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C(C)C)=C1N(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results