accessibility menu, dialog, popup

UserName

CH3OOHOH3C

CAS RN 2935-63-9

IUPAC Name: 2-(2,3-dimethylphenoxy)acetic acid
Synonyms: 2,3-dimethylphenoxyacetic acid
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: AVDBMBYECMTQJQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(OCC(O)=O)=C1C
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results