accessibility menu, dialog, popup

UserName

NH2HNOBr

CAS RN 29418-67-5

IUPAC Name: 2-bromobenzohydrazide
Synonyms: benzoylhydrazine, O-bromo
Molecular Weight (g/mol): 215.05
Molecular Formula: C7H7BrN2O
InChi Key: PQNLAYLOCZKPIY-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results