accessibility menu, dialog, popup

UserName

CAS RN 2942-06-5

ONONS

CAS RN 2942-06-5

IUPAC Name: 6-nitro-1,3-benzothiazole
Synonyms: 6-nitro-1,3-benzothiazole
Molecular Weight (g/mol): 180.18
Molecular Formula: C7H4N2O2S
InChi Key: QLUFBCVWKTWKBF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2N=CSC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results