missing translation for 'onlineSavingsMsg'
Learn More

6-Nitrobenzothiazole, 98+%

Product Code. 11493354
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493354 50 g 50g
11344639 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493354 Supplier Thermo Scientific Alfa Aesar Supplier No. A10634.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2942-06-5 | C7H4N2O2S | 180.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2942-06-5
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.18
MDL Number MFCD00014569
InChI Key QLUFBCVWKTWKBF-UHFFFAOYSA-N
IUPAC Name 6-nitro-1,3-benzothiazole

Specifications

Melting Point 173°C to 177°C
Quantity 50 g
Beilstein 7824
Synonym 6-nitrobenzothiazole, 6-nitrobenzo d thiazole, benzothiazole, 6-nitro, 6-nitro-benzothiazole, pubchem21845, acmc-1ciqi, #, benzothiazole, 6-nitro-6ci,7ci,8ci,9ci
SMILES C1=CC2=C(C=C1[N+](=O)[O-])SC=N2
PubChem CID 76256
Formula Weight 180.18
Percent Purity ≥98%
Chemical Name or Material 6-Nitrobenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.