accessibility menu, dialog, popup

UserName

ONONHN

CAS RN 2942-42-9

IUPAC Name: 7-nitro-1H-indazole
Synonyms: 7-nitroindazole
Molecular Weight (g/mol): 163.14
Molecular Formula: C7H5N3O2
InChi Key: PQCAUHUKTBHUSA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2NN=CC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results