missing translation for 'onlineSavingsMsg'
Learn More

7-Nitro-1H-indazole, 98%

Product Code. 11372127
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372127 25 g 25g
11352127 1 g 1g
11362127 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11372127 Supplier Thermo Scientific Alfa Aesar Supplier No. L07970.14
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2942-42-9 | C7H5N3O2 | 163.136 g/mol

7-nitroindazole is a specific inhibitor of neural nitric oxide synthase (nNOS), an enzyme that converts arginine to citrulline and nitric oxide (NO) in neuronal tissue. NO is a key player in the hippocampal long-term potentiation (LTP) and in general cell signaling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-nitroindazole is a specific inhibitor of neural nitric oxide synthase (nNOS), an enzyme that converts arginine to citrulline and nitric oxide (NO) in neuronal tissue. NO is a key player in the hippocampal long-term potentiation (LTP) and in general cell signaling.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2942-42-9
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.136
MDL Number MFCD00022789
InChI Key PQCAUHUKTBHUSA-UHFFFAOYSA-N
Synonym 7-nitroindazole, 1h-indazole, 7-nitro, 7-ni, 7-nitro-indazole, unii-ux0n37cmvh, ccris 3309, 2h-indazole,7-nitro, ux0n37cmvh, 7ni
PubChem CID 1893
IUPAC Name 7-nitro-1H-indazole
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])NN=C2

Specifications

Melting Point 184°C to 189°C
Quantity 25 g
Beilstein 6809
Solubility Information Insoluble in water.
Formula Weight 163.14
Percent Purity 98%
Chemical Name or Material 7-Nitro-1H-indazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.