accessibility menu, dialog, popup

UserName

CAS RN 295320-25-1

IUPAC Name: benzyl 2-(3-hydroxyphenyl)acetate
Synonyms: benzyl 2-(3-hydroxyphenyl)acetate
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: ALFOBMRIXXPJLQ-UHFFFAOYSA-N
SMILES: OC1=CC=CC(CC(=O)OCC2=CC=CC=C2)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results