CAS RN 29668-12-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethylene Terephthalate Cyclic Heptamer, TRC
CAS: 29668-12-0 Molecular Formula: C70 H56 O28 Molecular Weight (g/mol): 1345.177 Synonym: 3,6,13,16,23,26,33,36,43,46,53,56,63,66-Tetradecaoxaoctacyclo[66.2.2.28,11.218,21.228,31.238,41.248,51.258,61]tetraoctaconta-8,10,18,20,28,30,38,40,48,50,58,60,68,70,71,73,75,77,79,81,83-heneicosaene-2,7,12,17,22,27,32,37,42,47,52,57,62,67-tetradecone,Terephthalic Acid, Heptamol. Cyclic Ethylene Ester,Ethylene Glycol, Heptamol. Cyclic Terephthalate SMILES: O=C1OCCOC(=O)C2=CC=C(C=C2)C(=O)OCCOC(=O)C3=CC=C(C=C3)C(=O)OCCOC(=O)C4=CC=C(C=C4)C(=O)OCCOC(=O)C5=CC=C(C=C5)C(=O)OCCOC(=O)C6=CC=C(C=C6)C(=O)OCCOC(=O)C7=CC=C(C=C7)C(=O)OCCOC(=O)C8=CC=C1C=C8