CAS RN 29681-42-3
CAS RN 29681-42-3
IUPAC Name:
methyl 4-bromopyridine-2-carboxylate
Synonyms:
methyl 4-bromopyridine-2-carboxylate
Molecular Weight (g/mol):
216.03
Molecular Formula:
C7H6BrNO2
InChi Key:
JZFLATQBIPILFS-UHFFFAOYSA-N
SMILES:
COC(=O)C1=NC=CC(Br)=C1
1
–
1
of
1
results
1
Methyl 4-bromopyridine-2-carboxylate, 97+%
CAS: 29681-42-3 Molecular Formula: C7H6BrNO2 Molecular Weight (g/mol): 216.034 MDL Number: MFCD07374897 InChI Key: JZFLATQBIPILFS-UHFFFAOYSA-N PubChem CID: 12828947 IUPAC Name: methyl 4-bromopyridine-2-carboxylate SMILES: COC(=O)C1=NC=CC(=C1)Br