missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromopyridine-2-carboxylate, 97+%

Product Code. 15491479
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15491479

Brand: Thermo Scientific Alfa Aesar H64327.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29681-42-3 | C7H6BrNO2 | 216.034 g/mol

It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29681-42-3
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD07374897
InChI Key JZFLATQBIPILFS-UHFFFAOYSA-N
PubChem CID 12828947
IUPAC Name methyl 4-bromopyridine-2-carboxylate
SMILES COC(=O)C1=NC=CC(=C1)Br

Specifications

Melting Point 54°C to 56°C
Quantity 25 g
Beilstein 473047
Synonym methyl 4-bromopicolinate, 4-bromo-pyridine-2-carboxylic acid methyl ester, methyl 4-bromo-2-pyridinecarboxylate, methyl4-bromopicolinate, 2-pyridinecarboxylic acid, 4-bromo-, methyl ester, 4-bromopyridine-2-carboxylic acid methyl ester, abbypharma ap-10-5912, pubchem12733, acmc-209hag, methyl 4-bromo-pyridine-2-carboxylate
Solubility Information Slightly soluble in water.
Formula Weight 216.03
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material Methyl 4-bromopyridine-2-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.