accessibility menu, dialog, popup

UserName

SNNOOOOH3CCH3CH3CH3

CAS RN 29684-56-8

IUPAC Name: triethyl({[(methoxycarbonyl)imino](oxido)oxo-λ⁶-sulfanyl})azanium
Synonyms: burgess reagent
Molecular Weight (g/mol): 238.3
Molecular Formula: C8H18N2O4S
InChi Key: YSHOWEKUVWPFNR-UHFFFAOYSA-N
SMILES: CC[N+](CC)(CC)S([O-])(=O)=NC(=O)OC
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)