CAS RN 2973-75-3
IUPAC Name:
2,3-dibromo-4-hydroxy-5-methoxybenzaldehyde
Synonyms:
2,3-dibromo-4-hydroxy-5-methoxybenzaldehyde
Molecular Weight (g/mol):
309.94
Molecular Formula:
C8H6Br2O3
InChi Key:
WKLKGSHBXNPUDU-UHFFFAOYSA-N
SMILES:
COC1=CC(C=O)=C(Br)C(Br)=C1O
1
–
1
of
1
results
1
2,3-Dibromo-4-hydroxy-5-methoxybenzaldehyde, 98%
CAS: 2973-75-3 Molecular Formula: C8H6Br2O3 Molecular Weight (g/mol): 309.94 MDL Number: MFCD00016978 InChI Key: WKLKGSHBXNPUDU-UHFFFAOYSA-N Synonym: 5,6-dibromovanillin,acmc-1chm2,2,3-dibromo-5-methoxy-4-hydroxybenzaldehyde,2,3-bis bromanyl-5-methoxy-4-oxidanyl-benzaldehyde PubChem CID: 520452 IUPAC Name: 2,3-dibromo-4-hydroxy-5-methoxybenzaldehyde SMILES: COC1=CC(C=O)=C(Br)C(Br)=C1O