CAS RN 29748-10-5
CAS RN 29748-10-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Loganetin, MedChemExpress
MedChemExpress Loganetin is a non-toxic natural product that may be applied in the antibacterial drug development for treating multidrug-resistant Gram negative infections.