missing translation for 'onlineSavingsMsg'
Learn More

Loganetin, MedChemExpress

Product Code. 16481728
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481728 5 mg 5mg
16479298 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481728 Supplier Medchem Express Supplier No. HYN3373

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Loganetin is a non-toxic natural product that may be applied in the antibacterial drug development for treating multidrug-resistant Gram negative infections.

Loganetin is a non-toxic natural product that may be applied in the antibacterial drug development for treating multidrug-resistant Gram negative infections.

Specifications

Chemical Name or Material Loganetin
CAS 29748-10-5
Color Colorless
Purity Grade Notes Research
Molecular Formula C11H16O5
Quantity 5 mg
Solubility Information DMSO : 250 mg/mL (1095.34 mM; Need ultrasonic)
SMILES COC(C1=CO[C@@H](O)[C@@]2([H])[C@]1([H])C[C@H](O)[C@@H]2C)=O
Molecular Weight (g/mol) 228.24
Formula Weight 228.24
Percent Purity 98.19%
Grade Research
Physical Form Oil
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.