accessibility menu, dialog, popup

UserName

HOOO

CAS RN 2976-75-2

IUPAC Name: 2-(naphthalen-1-yloxy)acetic acid
Synonyms: naphthyloxyacetic acid 1-noa cpd 1-naphthyloxyacetic acid 1-naphthoxyacetic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C12H10O3
InChi Key: GHRYSOFWKRRLMI-UHFFFAOYSA-N
SMILES: OC(=O)COC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results