accessibility menu, dialog, popup

UserName

CH3CH3NH2HC

CAS RN 2978-58-7

IUPAC Name: 2-methylbut-3-yn-2-amine
Synonyms: 3-butyn-2-amine, 2-methyl-
Molecular Weight (g/mol): 83.13
Molecular Formula: C5H9N
InChi Key: VUGCBIWQHSRQBZ-UHFFFAOYSA-N
SMILES: CC(C)(N)C#C
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)