missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylpropargylamine, 95%

Product Code. 15470268
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470268 5 g 5g
15480268 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470268 Supplier Thermo Scientific Alfa Aesar Supplier No. H53448.06
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2978-58-7 | C5H9N | 83.134 g/mol

1,1-Dimethylpropargylamine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Dimethylpropargylamine is used as an organic chemical synthesis intermediate.

Solubility
Very soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2978-58-7
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00008052
InChI Key VUGCBIWQHSRQBZ-UHFFFAOYSA-N
Synonym 1,1-dimethylpropargylamine, 3-amino-3-methyl-1-butyne, 3-butyn-2-amine, 2-methyl, 1,1-dimethylprop-2-ynylamine, 2-methyl-3-butyn-2-amine, 1,1-dimethyl-prop-2-ynylamine, 1,1-dimethylpropynylamine, 1,1-dimethyl-2-propynylamine, 1,1-dimethylprop-3-ynylamine, 3-amino-3-methylbutyne
PubChem CID 76319
IUPAC Name 2-methylbut-3-yn-2-amine
SMILES CC(C)(C#C)N

Specifications

Density 0.79
Boiling Point 79°C to 80°C
Flash Point 2°C (36°F)
Refractive Index 1.4237
Quantity 5 g
UN Number UN2733
Sensitivity Air Sensitive
Solubility Information Very soluble in water.
Formula Weight 83.13
Percent Purity 95%
Chemical Name or Material 1,1-Dimethylpropargylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.