CAS RN 29906-67-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Methyl-5-nitro-1H-indole, TRC
CAS: 29906-67-0 Molecular Formula: C9H8N2O2 Molecular Weight (g/mol): 176.17 Synonym: 1-Methyl-5-nitroindole,5-Nitro-1-methylindole,N-Methyl-5-nitroindole IUPAC Name: 1-methyl-5-nitroindole SMILES: Cn1ccc2cc(ccc12)[N+](=O)[O-]