missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-nitro-1H-indole, TRC

Product Code. 30480488
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480488 25 g 25g
30464905 1 g 1g
30467480 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480488 Supplier Toronto Research Chemicals Supplier No. TRCM32097025G
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29906-67-0
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.17
Synonym 1-Methyl-5-nitroindole, 5-Nitro-1-methylindole, N-Methyl-5-nitroindole
IUPAC Name 1-methyl-5-nitroindole
SMILES Cn1ccc2cc(ccc12)[N+](=O)[O-]

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H8N2O2/c1-10-5-4-7-6-8(11(12)13)2-3-9(7)10/h2-6H,1H3
Formula Weight 176.0586
Chemical Name or Material 1-Methyl-5-nitro-1H-indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.