accessibility menu, dialog, popup

UserName

CH3OPOCH3OCH3

CAS RN 29949-84-6

IUPAC Name: tris(3-methoxyphenyl)phosphane
Synonyms: tris(3-methoxyphenyl)phosphane
Molecular Weight (g/mol): 352.37
Molecular Formula: C21H21O3P
InChi Key: CCXTYQMZVYIQRP-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1)P(C1=CC(OC)=CC=C1)C1=CC=CC(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results